Methyl-1-(2-nitro-4-(trifluormethyl)phenyl)cyclopropancarboxylat
Lieferant: COMBI-BLOCKS
AN-1189-1G
AN-1189-5G
COBBAN-1189-1GEA
157
EUR
COBBAN-1189-1G
COBBAN-1189-5G
Methyl-1-(2-nitro-4-(trifluormethyl)phenyl)cyclopropancarboxylat
Methyl-1-(2-nitro-4-(trifluormethyl)phenyl)cyclopropancarboxylat
Formel:
C₁₂H₁₀F₃NO₄ |
MDL:
MFCD09878310 CAS-Nummer: 951885-67-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...