1-(4-Fluorphenyl)cyclopropancarbonsäure

Lieferant: COMBI-BLOCKS

HC-2894-5G HC-2894-1G
COBBHC-2894-5GEA 308 EUR
COBBHC-2894-5G COBBHC-2894-1G
1-(4-Fluorphenyl)cyclopropancarbonsäure
1-(4-Fluorophenyl)cyclopropanecarboxylic acid
Formel: C₁₀H₉FO₂
Molecular Weight: 180,1800 g/mol
MDL: MFCD06208777
CAS-Nummer: 773100-29-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR