4-Brom-1-(2-tetrahydropyranyl)-1H-pyrazol

Lieferant: COMBI-BLOCKS

Synonyme: 4-Bromo-1-(tetrahydro-2H-pyran-2-yl)-1H-pyrazole, 4-Bromo-1-(tetrahydro-2H-pyran-2-yl-1H)-pyrazole

HI-1505-1G HI-1505-5G
COBBHI-1505-1GEA 35.9 EUR
COBBHI-1505-1G COBBHI-1505-5G
4-Brom-1-(2-tetrahydropyranyl)-1H-pyrazol
4-Brom-1-(2-tetrahydropyranyl)-1H-pyrazol
Formel: C₈H₁₁BrN₂O
Molecular Weight: 231.0900 g/mol
MDL: MFCD12026052
CAS-Nummer: 82099-98-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR