Ethyl-3,4,5-trimethylpyrrol-2-carboxylat ≥98%

Lieferant: Thermo Scientific

A13814.03 A13814.06
A13814.03EA 40 EUR
A13814.03 A13814.06
Ethyl-3,4,5-trimethylpyrrol-2-carboxylat ≥98%
Ethyl-3,4,5-trimethylpyrrol-2-carboxylat

Purity: 98% CAS No.: 2199-46-4 Molecular Formula: C10H15NO2 Formula Weight: 181.24 Melting Point: 124-128° Beilstein Reference: 132613

Formel: C₁₀H₁₅NO₂
Molecular Weight: 181,24 g/mol
Schmelzpunkt: 124…128 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00051948
CAS-Nummer: 2199-46-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR