3-(3-(Ethoxycarbonyl)piperidin-1-carbonyl)phenylboronsäurepinakolester

Lieferant: COMBI-BLOCKS

Synonyme: Ethyl 1-(3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzoyl)piperidine-3-carboxylate

PN-8798-5G PN-8798-1G
COBBPN-8798-5GEA 0 EUR
COBBPN-8798-5G COBBPN-8798-1G
3-(3-(Ethoxycarbonyl)piperidin-1-carbonyl)phenylboronsäurepinakolester
3-(3-(Ethoxycarbonyl)piperidin-1-carbonyl)phenylboronsäurepinakolester
Formel: C₂₁H₃₀BNO₅
MDL: MFCD09266178
CAS-Nummer: 1073355-04-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR