2-(Piperidin-1-ylmethyl)phenylboronsäure
Lieferant: COMBI-BLOCKS
Synonyme:
(2-(Piperidin-1-ylmethyl)phenyl)boronic acid
BB-5437-1G
BB-5437-250M
COBBBB-5437-1GEA
884
EUR
COBBBB-5437-1G
COBBBB-5437-250M
2-(Piperidin-1-ylmethyl)phenylboronsäure
2-(Piperidin-1-ylmethyl)phenylboronsäure
Formel:
C₁₂H₁₈BNO₂ Molecular Weight: 219.0900 g/mol |
MDL:
MFCD06801661 CAS-Nummer: 878289-33-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...