3-(Piperidin-1-carbonyl)phenylboronsäure

Lieferant: COMBI-BLOCKS

BB-3053-5G BB-3053-25G BB-3053-1G
COBBBB-3053-5GEA 251 EUR
COBBBB-3053-5G COBBBB-3053-25G COBBBB-3053-1G
3-(Piperidin-1-carbonyl)phenylboronsäure
3-(Piperidin-1-carbonyl)phenylboronsäure
Formel: C₁₂H₁₆BNO₃
Molecular Weight: 233.0713 g/mol
MDL: MFCD04115686
CAS-Nummer: 850568-34-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR