4-(Piperidin-1-carbonyl)phenylboronsäure
Lieferant: COMBI-BLOCKS
Synonyme:
4-[(Piperidin-1-yl)carbonyl]benzeneboronic acid
BB-3003-1G
BB-3003-5G
COBBBB-3003-1GEA
61.9
EUR
COBBBB-3003-1G
COBBBB-3003-5G
4-(Piperidin-1-carbonyl)phenylboronsäure
4-(Piperidin-1-carbonyl)phenylboronsäure
Formel:
C₁₂H₁₆BNO₃ Molecular Weight: 233.0713 g/mol |
MDL:
MFCD03411951 CAS-Nummer: 389621-83-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...