4-(4-Carboxypiperidin-1-carbonyl)phenylboronsäure
Lieferant: COMBI-BLOCKS
Synonyme:
1-(4-Boronobenzoyl)piperidine-4-carboxylic acid, 4-(1-Carbonyl-4-carboxypiperidine)benzeneboronic acid
BB-2782-5G
BB-2782-1G
COBBBB-2782-5GEA
931
EUR
COBBBB-2782-5G
COBBBB-2782-1G
4-(4-Carboxypiperidin-1-carbonyl)phenylboronsäure
4-(4-Carboxypiperidin-1-carbonyl)phenylboronsäure
Formel:
C₁₃H₁₆BNO₅ Molecular Weight: 277,0800 g/mol |
MDL:
MFCD07363784 CAS-Nummer: 850593-02-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...