3-Nitro-5-(piperidin-1-carbonyl)phenylboronsäure

Lieferant: COMBI-BLOCKS

BB-2824-5G BB-2824-1G
COBBBB-2824-5GEA 303 EUR
COBBBB-2824-5G COBBBB-2824-1G
3-Nitro-5-(piperidin-1-carbonyl)phenylboronsäure
3-Nitro-5-(piperidin-1-carbonyl)phenylboronsäure
Formel: C₁₂H₁₅BN₂O₅
MDL: MFCD07783871
CAS-Nummer: 871332-78-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR