3-Nitro-5-(1-pyrrolidinylcarbonyl)phenylboronsäure

Lieferant: COMBI-BLOCKS

Synonyme: (3-Nitro-5-(pyrrolidine-1-carbonyl)phenyl)boronic acid

BB-2828-5G BB-2828-1G
COBBBB-2828-5GEA 692 EUR
COBBBB-2828-5G COBBBB-2828-1G
3-Nitro-5-(1-pyrrolidinylcarbonyl)phenylboronsäure
3-Nitro-5-(1-pyrrolidinylcarbonyl)phenylboronsäure
Formel: C₁₁H₁₃BN₂O₅
Molecular Weight: 264.0400 g/mol
MDL: MFCD07783873
CAS-Nummer: 871332-81-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR