3-(4-Oxopiperidin-1-carbonyl)phenylboronsäure

Lieferant: COMBI-BLOCKS

BB-3076-5G BB-3076-1G
COBBBB-3076-5GEA 918 EUR
COBBBB-3076-5G COBBBB-3076-1G
3-(4-Oxopiperidin-1-carbonyl)phenylboronsäure
3-(4-Oxopiperidin-1-carbonyl)phenylboronsäure
Formel: C₁₂H₁₄BNO₄
MDL: MFCD04115707
CAS-Nummer: 850567-32-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR