3-(3-(Ethoxycarbonyl)piperidin-1-carbonyl)phenylboronsäure

Lieferant: COMBI-BLOCKS

Synonyme: (3-(3-(Ethoxycarbonyl)piperidine-1-carbonyl)phenyl)boronic acid

BB-8798-5G BB-8798-1G
COBBBB-8798-5GEA 595 EUR
COBBBB-8798-5G COBBBB-8798-1G
3-(3-(Ethoxycarbonyl)piperidin-1-carbonyl)phenylboronsäure
3-(3-(Ethoxycarbonyl)piperidin-1-carbonyl)phenylboronsäure
Formel: C₁₅H₂₀BNO₅
Molecular Weight: 305.1300 g/mol
MDL: MFCD12756423
CAS-Nummer: 1218790-81-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR