4-(3-(Ethoxycarbonyl)piperidin-1-carbonyl)phenylboronsäure

Lieferant: COMBI-BLOCKS

Synonyme: (4-(3-(Ethoxycarbonyl)piperidine-1-carbonyl)phenyl)boronic acid

BB-8657-5G BB-8657-1G
COBBBB-8657-5GEA 884 EUR
COBBBB-8657-5G COBBBB-8657-1G
4-(3-(Ethoxycarbonyl)piperidin-1-carbonyl)phenylboronsäure
4-(3-(Ethoxycarbonyl)piperidin-1-carbonyl)phenylboronsäure
Formel: C₁₅H₂₀BNO₅
Molecular Weight: 305.1300 g/mol
MDL: MFCD11504863
CAS-Nummer: 1150114-74-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR