4-Chlor-3-(piperidin-1-carbonyl)phenylboronsäure

Lieferant: COMBI-BLOCKS

Synonyme: (4-Chloro-3-(piperidine-1-carbonyl)phenyl)boronic acid

BB-2814-5G BB-2814-1G
COBBBB-2814-5GEA 451 EUR
COBBBB-2814-5G COBBBB-2814-1G
4-Chlor-3-(piperidin-1-carbonyl)phenylboronsäure
4-Chlor-3-(piperidin-1-carbonyl)phenylboronsäure
Formel: C₁₂H₁₅BClNO₃
Molecular Weight: 267,5200 g/mol
MDL: MFCD07783862
CAS-Nummer: 871332-70-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR