3-Chlor-5-(piperidin-1-carbonyl)phenylboronsäure

Lieferant: COMBI-BLOCKS

Synonyme: (3-Chloro-5-(piperidine-1-carbonyl)phenyl)boronic acid, 3-Chloro-5-(piperidin-1-carbonyl)phenylboronic acid

BB-3243-5G BB-3243-1G
COBBBB-3243-5GEA 895 EUR
COBBBB-3243-5G COBBBB-3243-1G
3-Chlor-5-(piperidin-1-carbonyl)phenylboronsäure
3-Chlor-5-(piperidin-1-carbonyl)phenylboronsäure
Formel: C₁₂H₁₅BClNO₃
Molecular Weight: 267,5200 g/mol
MDL: MFCD09800859
CAS-Nummer: 957120-47-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR