4-Chlor-3-(1-pyrrolidinylcarbonyl)phenylboronsäure
Lieferant: COMBI-BLOCKS
Synonyme:
(4-Chloro-3-(pyrrolidine-1-carbonyl)phenyl)boronic acid
BB-2819-5G
BB-2819-1G
COBBBB-2819-5GEA
0
EUR
COBBBB-2819-5G
COBBBB-2819-1G
4-Chlor-3-(1-pyrrolidinylcarbonyl)phenylboronsäure
4-Chlor-3-(1-pyrrolidinylcarbonyl)phenylboronsäure
Formel:
C₁₁H₁₃BClNO₃ |
MDL:
MFCD07783867 CAS-Nummer: 871332-75-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...