3-Chlor-4-(piperidin-1-carbonyl)phenylboronsäure
Lieferant: COMBI-BLOCKS
Synonyme:
3-Chloro-4-(1-piperidinylcarbonyl)benzeneboronic acid, (3-Chloro-4-(piperidine-1-carbonyl)phenyl)boronic acid
BB-2770-5G
BB-2770-1G
COBBBB-2770-5GEA
1580
EUR
COBBBB-2770-5G
COBBBB-2770-1G
3-Chlor-4-(piperidin-1-carbonyl)phenylboronsäure
3-Chlor-4-(piperidin-1-carbonyl)phenylboronsäure
Formel:
C₁₂H₁₅BClNO₃ Molecular Weight: 267.52 g/mol |
MDL:
MFCD07363775 CAS-Nummer: 850589-50-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...