3-Chlor-4-(1-pyrrolidinylcarbonyl)phenylboronsäure

Lieferant: COMBI-BLOCKS

Synonyme: 3-Chloro-4-(1-pyrrolidinylcarbonyl)benzeneboronic acid, 3-Chloro-4-(pyrrolidinyl-1-carbonyl)phenylboronic acid

BB-2771-5G BB-2771-1G
COBBBB-2771-5GEA 0 EUR
COBBBB-2771-5G COBBBB-2771-1G
3-Chlor-4-(1-pyrrolidinylcarbonyl)phenylboronsäure
3-Chlor-4-(1-pyrrolidinylcarbonyl)phenylboronsäure
Formel: C₁₁H₁₃BClNO₃
MDL: MFCD07363776
CAS-Nummer: 850589-51-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR