4-Fluor-3-(piperidin-1-carbonyl)phenylboronsäure
Lieferant: COMBI-BLOCKS
BB-2873-1G
BB-2873-5G
COBBBB-2873-1GEA
0
EUR
COBBBB-2873-1G
COBBBB-2873-5G
4-Fluor-3-(piperidin-1-carbonyl)phenylboronsäure
4-Fluor-3-(piperidin-1-carbonyl)phenylboronsäure
Formel:
C₁₂H₁₅BFNO₃ |
MDL:
MFCD08235049 CAS-Nummer: 874219-30-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...