3-Fluor-5-(piperidin-1-carbonyl)phenylboronsäure

Lieferant: COMBI-BLOCKS

Synonyme: (3-Fluoro-5-(piperidine-1-carbonyl)phenyl)boronic acid

BB-2858-5G BB-2858-1G
COBBBB-2858-5GEA 1430 EUR
COBBBB-2858-5G COBBBB-2858-1G
3-Fluor-5-(piperidin-1-carbonyl)phenylboronsäure
3-Fluor-5-(piperidin-1-carbonyl)phenylboronsäure
Formel: C₁₂H₁₅BFNO₃
Molecular Weight: 251,0600 g/mol
MDL: MFCD08235060
CAS-Nummer: 874219-43-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR