2-Fluor-5-(piperidin-1-carbonyl)phenylboronsäure

Lieferant: COMBI-BLOCKS

Synonyme: (2-Fluoro-5-(piperidine-1-carbonyl)phenyl)boronic acid, 2-Fluoro-5-(1-piperidinylcarbonyl)benzeneboronic acid

BB-3338-5G BB-3338-1G
COBBBB-3338-5GEA 0 EUR
COBBBB-3338-5G COBBBB-3338-1G
2-Fluor-5-(piperidin-1-carbonyl)phenylboronsäure
2-Fluor-5-(piperidin-1-carbonyl)phenylboronsäure
Formel: C₁₂H₁₅BFNO₃
Molecular Weight: 251.06 g/mol
MDL: MFCD08436068
CAS-Nummer: 874289-43-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR