4-Fluor-3-(1-pyrrolidinylcarbonyl)phenylboronsäure

Lieferant: COMBI-BLOCKS

Synonyme: 4-Fluoro-3-(1-pyrrolidinylcarbonyl)benzeneboronic acid, 4-Fluoro-3-(pyrrolidine-1-carbonyl)phenylboronic acid

BB-2874-5G BB-2874-1G
COBBBB-2874-5GEA 0 EUR
COBBBB-2874-5G COBBBB-2874-1G
4-Fluor-3-(1-pyrrolidinylcarbonyl)phenylboronsäure
4-Fluor-3-(1-pyrrolidinylcarbonyl)phenylboronsäure
Formel: C₁₁H₁₃BFNO₃
MDL: MFCD08235050
CAS-Nummer: 874219-31-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR