2-Fluor-5-(1-pyrrolidinylcarbonyl)phenylboronsäure

Lieferant: COMBI-BLOCKS

Synonyme: (2-Fluoro-5-(pyrrolidine-1-carbonyl)phenyl)boronic acid, 2-Fluoro-5-(pyrolidine-1-carbonyl)phenylboronic acid

BB-3337-5G BB-3337-1G
COBBBB-3337-5GEA 1690 EUR
COBBBB-3337-5G COBBBB-3337-1G
2-Fluor-5-(1-pyrrolidinylcarbonyl)phenylboronsäure
2-Fluor-5-(1-pyrrolidinylcarbonyl)phenylboronsäure
Formel: C₁₁H₁₃BFNO₃
Molecular Weight: 237.0400 g/mol
MDL: MFCD08436030
CAS-Nummer: 874289-42-6

Bestellen

TECHNISCHE DATEN

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR