3-Fluor-4-(1-pyrrolidinylcarbonyl)phenylboronsäure

Lieferant: COMBI-BLOCKS

Synonyme: 3-Fluoro-4-(pyrolidine-1-carbonyl)phenylboronic acid, (3-Fluoro-4-(pyrrolidine-1-carbonyl)phenyl)boronic acid

BB-3305-5G BB-3305-1G
COBBBB-3305-5GEA 0 EUR
COBBBB-3305-5G COBBBB-3305-1G
3-Fluor-4-(1-pyrrolidinylcarbonyl)phenylboronsäure
3-Fluor-4-(1-pyrrolidinylcarbonyl)phenylboronsäure
Formel: C₁₁H₁₃BFNO₃
MDL: MFCD08436024
CAS-Nummer: 874289-09-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR