3-Brom-4-nitrobenzotrifluorid
Lieferant: COMBI-BLOCKS
Synonyme:
2-Bromo-1-nitro-4-(trifluoromethyl)benzene
HC-6656-5G
HC-6656-1G
COBBHC-6656-5GEA
321
EUR
COBBHC-6656-5G
COBBHC-6656-1G
3-Brom-4-nitrobenzotrifluorid
3-Brom-4-nitrobenzotrifluorid
Formel:
C₇H₃BrF₃NO₂ Molecular Weight: 270,01 g/mol |
MDL:
MFCD09998160 CAS-Nummer: 132839-58-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...