Chlorodimethyloctadecylsilan

Lieferant: CHEMPUR SILANE

SILS12175.25 SILS12175.100
SILNSILS12175.25EA 0 EUR
SILNSILS12175.25 SILNSILS12175.100
Chlorodimethyloctadecylsilan
Chlorodimethyloctadecylsilan
Formel: C₂₀H₄₃ClSi
Molecular Weight: 347.10 g/mol
MDL: MFCD00000504
CAS-Nummer: 18643-08-8
EINECS: 242-472-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR