1-tert-Butyl-4-piperidon

Lieferant: COMBI-BLOCKS

Synonyme: 1-(tert-Butyl)piperidin-4-one

HC-6270-1G HC-6270-5G
COBBHC-6270-1GEA 174 EUR
COBBHC-6270-1G COBBHC-6270-5G
1-tert-Butyl-4-piperidon
1-tert-Butyl-4-piperidon
Formel: C₉H₁₇NO
Molecular Weight: 155.24 g/mol
MDL: MFCD03411608
CAS-Nummer: 1465-76-5
EINECS: 226-339-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR