1-Isopropyl-4-piperidon
Lieferant: COMBI-BLOCKS
HC-3168-100G
HC-3168-25G
HC-3168-5G
COBBHC-3168-100GEA
470
EUR
COBBHC-3168-100G
COBBHC-3168-25G
COBBHC-3168-5G
1-Isopropyl-4-piperidon
1-Isopropyl-4-piperidon
Formel:
C₈H₁₅NO Molecular Weight: 141,22 g/mol |
MDL:
MFCD00038035 CAS-Nummer: 5355-68-0 EINECS: 226-339-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...