4-Methyloctansäure, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ Qualitätsstufe = MQ300

W357502-100G-K W357502-1KG-K W357502-25G-K
SIALW357502-100G-KEA 624 EUR
SIALW357502-100G-K SIALW357502-1KG-K SIALW357502-25G-K
4-Methyloctansäure, Sigma-Aldrich®
4-Methylcaprylsäure
Formel: C₉H₁₈O₂
MDL: MFCD00051938
CAS-Nummer: 54947-74-9
EINECS: 259-404-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR