10-Hydroxydecansäure, Sigma-Aldrich®

Lieferant: Merck

379700-500MG 379700-5G
SIAL379700-500MGEA 88.4 EUR
SIAL379700-500MG SIAL379700-5G
10-Hydroxydecansäure, Sigma-Aldrich®
10-Hydroxydecanoic acid
Formel: C₁₀H₂₀O₃
Molecular Weight: 188,26 g/mol
MDL: MFCD00010510
CAS-Nummer: 1679-53-4
EINECS: 216-848-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR