1-(2-Phenylethyl)piperazin ≥98%
Lieferant: Thermo Scientific
|
Gefahr
|
B25158.09
B25158.18
B25158.09EA
41.5
EUR
B25158.09
B25158.18
1-(2-Phenylethyl)piperazin ≥98%
1-(2-Phenylethyl)piperazin
Purity: 98%
CAS No.: 5321-49-3
Molecular Formula: C12H18N2
Formula Weight: 190.29
Flash Point:
Formel:
C₁₂H₁₈N₂ Molecular Weight: 190,29 g/mol Siedepunkt: 276 °C Dichte: 1,01 Flammpunkt: >110 °C (230 °F) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00040740 CAS-Nummer: 5321-49-3 EINECS: 226-186-5 ADR: 8,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...