1-Brom-2-methyl-4-(trifluormethoxy)benzol
Lieferant: COMBI-BLOCKS
Synonyme:
2-Bromo-5-(trifluoromethoxy)toluene
OT-0529-5G
OT-0529-1G
COBBOT-0529-5GEA
149
EUR
COBBOT-0529-5G
COBBOT-0529-1G
1-Brom-2-methyl-4-(trifluormethoxy)benzol
1-Bromo-2-methyl-4-(trifluoromethoxy)benzene
Formel:
C₈H₆BrF₃O Molecular Weight: 255,0319 g/mol |
MDL:
MFCD01631545 CAS-Nummer: 261951-96-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...