2-Brom-6-fluor-3-methylanisol

Lieferant: COMBI-BLOCKS

Synonyme: 2-Bromo-4-fluoro-3-methoxy-1-methylbenzene

OT-1231-5G OT-1231-1G
COBBOT-1231-5GEA 884 EUR
COBBOT-1231-5G COBBOT-1231-1G
2-Brom-6-fluor-3-methylanisol
2-Brom-6-fluor-3-methylanisol
Formel: C₈H₈BrFO
MDL: MFCD16295102
CAS-Nummer: 1226808-62-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR