3-Amino-2,4-dibrom-6-methylpyridin

Lieferant: COMBI-BLOCKS

PY-7959-1G PY-7959-250M
COBBPY-7959-1GEA 692 EUR
COBBPY-7959-1G COBBPY-7959-250M
3-Amino-2,4-dibrom-6-methylpyridin
3-Amino-2,4-dibromo-6-methylpyridine
Formel: C₆H₆Br₂N₂
MDL: MFCD07774092
CAS-Nummer: 706789-26-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR