4-Chlor-2-pyridincarbonitril
Lieferant: COMBI-BLOCKS
CA-4131-5G
CA-4131-25G
CA-4131-1G
COBBCA-4131-5GEA
76.3
EUR
COBBCA-4131-5G
COBBCA-4131-25G
COBBCA-4131-1G
4-Chlor-2-pyridincarbonitril
4-Chlor-2-pyridincarbonitril
Formel:
C₆H₃ClN₂ Molecular Weight: 138,5500 g/mol |
MDL:
MFCD06009833 CAS-Nummer: 19235-89-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...