4-Chlor-7-methylisatin
Lieferant: SIA ENAMINE
Synonyme:
4-Chloro-7-methylIsatin, 4-Chloro-7-methylindoline-2,3-dione
EN300-60309
EN300-60309.10
ENAMEN300-60309EA
0
EUR
ENAMEN300-60309
ENAMEN300-60309.10
4-Chlor-7-methylisatin
4-Chlor-7-methylisatin
Formel:
C₉H₆ClNO₂ Molecular Weight: 195.6024 g/mol |
MDL:
MFCD00047216 CAS-Nummer: 61258-72-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...