3,4-Dichlor-o-toluidin

Lieferant: COMBI-BLOCKS

Synonyme: 3,4-Dichloro-2-methylaniline

AN-1155-5G AN-1155-1G
COBBAN-1155-5GEA 595 EUR
COBBAN-1155-5G COBBAN-1155-1G
3,4-Dichlor-o-toluidin
3,4-Dichlor-o-toluidin
Formel: C₇H₇Cl₂N
MDL: MFCD11855814
CAS-Nummer: 62077-25-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR