4,5-Dichlor-o-toluidin

Lieferant: COMBI-BLOCKS

Synonyme: 4,5-Dichloro-2-methylaniline

AN-1418-5G AN-1418-1G
COBBAN-1418-5GEA 595 EUR
COBBAN-1418-5G COBBAN-1418-1G
4,5-Dichlor-o-toluidin
4,5-Dichlor-o-toluidin
Formel: C₇H₇Cl₂N
Molecular Weight: 176,0432 g/mol
MDL: MFCD11855825
CAS-Nummer: 2387-08-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR