2-Amino-4,6-dichlorphenol
Lieferant: Thermo Scientific
Synonyme:
3,5-Dichloro-6-hydroxyaniline
BTB04021.250
BTB04021.1
MAYBBTB04021.250EA
0
EUR
MAYBBTB04021.250
MAYBBTB04021.1
2-Amino-4,6-dichlorphenol
2-Amino-4,6-dichlorphenol
Formel:
C₆H₅Cl₂NO Molecular Weight: 178,01 g/mol |
MDL:
MFCD00035766 CAS-Nummer: 527-62-8 EINECS: 208-421-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...