2-Cyan-4'-methylbiphenyl ≥98%
Lieferant: Thermo Scientific
|
Gefahr
|
Synonyme:
4'-Methyl-[1,1'-biphenyl]-2-carbonitrile
B21273.06
B21273.14
B21273.22
B21273.06EA
48.7
EUR
B21273.06
B21273.14
B21273.22
2-Cyan-4'-methylbiphenyl ≥98%
2-Cyan-4'-methylbiphenyl
Purity: 98+%
CAS No.: 114772-53-1
Molecular Formula: C14H11N
Formula Weight: 193.26
Melting Point: 49-53°
Boiling Point:
Formel:
C₁₄H₁₁N Molecular Weight: 193,26 g/mol Siedepunkt: >320 °C Schmelzpunkt: 49…53 °C Dichte: 1,170 Lagertemperatur: Raumtemperatur |
MDL:
MFCD00151805 CAS-Nummer: 114772-53-1 EINECS: 422-310-9 ADR: 9,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...