2,2-Diphenylglycin ≥98%

Lieferant: Thermo Fisher Scientific

Synonyme: alpha,alpha-Diphenylglycine, Diphenylglycin, 2-Amino-2,2-diphenylacetic acid

B22650.06 B22650.14
B22650.06EA 72.6 EUR
B22650.06 B22650.14
2,2-Diphenylglycin ≥98%
2,2-Diphenylglycin

Purity: 98% CAS No.: 3060-50-2 Molecular Formula: C14H13NO2 Formula Weight: 227.26 Melting Point: ca 246° dec. MDL No.: MFCD00008048

Formel: C₁₄H₁₃NO₂
Schmelzpunkt: ∼ 246 °C (Zersetzung)
Lagertemperatur: Raumtemperatur
MDL: MFCD00008048
CAS-Nummer: 3060-50-2
EINECS: 221-299-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR