3-(3,5-Di-tert-butyl-4-hydroxyphenyl)propionsäure

Lieferant: BIONET RESEARCH

Synonyme: 3,5-Di-tert-butyl-4-hydroxyphenylpropionic acid

8X-0734.500 8X-0734.1 8X-0734.5
BIOT8X-0734.500EA 0 EUR
BIOT8X-0734.500 BIOT8X-0734.1 BIOT8X-0734.5
3-(3,5-Di-tert-butyl-4-hydroxyphenyl)propionsäure
3-(3,5-Di-tert-butyl-4-hydroxyphenyl)propionsäure
Formel: C₁₇H₂₆O₃
MDL: MFCD00017519
CAS-Nummer: 20170-32-5
EINECS: 243-556-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR