3-(2-Phenoxyphenyl)propionsäure

Lieferant: BIONET RESEARCH

8X-0704.500 8X-0704.1 8X-0704.5 8X-0704.10
BIOT8X-0704.500EA 0 EUR
BIOT8X-0704.500 BIOT8X-0704.1 BIOT8X-0704.5 BIOT8X-0704.10
3-(2-Phenoxyphenyl)propionsäure
3-(2-Phenoxyphenyl)propionsäure
Formel: C₁₅H₁₄O₃
MDL: MFCD05663814
CAS-Nummer: 40492-92-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR