2-Fluor-4-(trifluormethyl)zimtsäure
Lieferant: FLUOROCHEM
007927-1G
007927-5G
FLUO007927-1GEA
0
EUR
FLUO007927-1G
FLUO007927-5G
2-Fluor-4-(trifluormethyl)zimtsäure
2-Fluor-4-(trifluormethyl)zimtsäure
Formel:
C₁₀H₆F₄O₂ |
MDL:
MFCD00236292 CAS-Nummer: 262608-88-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...