2-Fluor-5-(trifluormethyl)zimtsäure

Lieferant: FLUOROCHEM

007928-5G 007928-1G
FLUO007928-5GEA 0 EUR
FLUO007928-5G FLUO007928-1G
2-Fluor-5-(trifluormethyl)zimtsäure
2-Fluor-5-(trifluormethyl)zimtsäure
Formel: C₁₀H₆F₄O₂
MDL: MFCD00236293
CAS-Nummer: 247113-91-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR