3-Fluor-4-(trifluormethyl)zimtsäure
Lieferant: FLUOROCHEM
007930-1G
007930-5G
FLUO007930-1GEA
42.9
EUR
FLUO007930-1G
FLUO007930-5G
3-Fluor-4-(trifluormethyl)zimtsäure
3-Fluor-4-(trifluormethyl)zimtsäure
Formel:
C₁₀H₆F₄O₂ |
MDL:
MFCD00236295 CAS-Nummer: 231291-13-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...