3-Fluor-5-(trifluormethyl)zimtsäure
Lieferant: FLUOROCHEM
007931-1G
007931-5G
FLUO007931-1GEA
42.9
EUR
FLUO007931-1G
FLUO007931-5G
3-Fluor-5-(trifluormethyl)zimtsäure
3-Fluor-5-(trifluormethyl)zimtsäure
Formel:
C₁₀H₆F₄O₂ |
MDL:
MFCD00236296 CAS-Nummer: 575469-96-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...