4-Fluor-3-(trifluormethyl)zimtsäure
Lieferant: FLUOROCHEM
Synonyme:
3-(4-Fluoro-3-(trifluoromethyl)phenyl)acrylic acid
007933-5G
007933-1G
FLUO007933-5GEA
186
EUR
FLUO007933-5G
FLUO007933-1G
4-Fluor-3-(trifluormethyl)zimtsäure
4-Fluor-3-(trifluormethyl)zimtsäure
Formel:
C₁₀H₆F₄O₂ Molecular Weight: 234.15 g/mol |
MDL:
MFCD00236298 CAS-Nummer: 239463-90-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...