4-Ethoxy-2-fluor-1-nitrobenzol

Lieferant: COMBI-BLOCKS

AN-1027-5G AN-1027-25G AN-1027-1G
COBBAN-1027-5GEA 303 EUR
COBBAN-1027-5G COBBAN-1027-25G COBBAN-1027-1G
4-Ethoxy-2-fluor-1-nitrobenzol
4-Ethoxy-2-fluor-1-nitrobenzol
Formel: C₈H₈FNO₃
MDL: MFCD04115633
CAS-Nummer: 28987-48-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR